Thermo Scientific™ Capital Vial Univeral Test Tube Plug Caps
For plastic or glass test tubes
$113.82 - $113.82
Specifications
For Use With Fit tubes with 12 to 16mm diameter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
21-235-720 Thermo Scientific™
M995135
White Pack of 1000 Each for $113.82
Specifications
Description
Innovative flanged-thread design allows cap to descend into the tube until a friction fit is secure, and plug is stationary.
  • Fit tubes with 12 to 16mm diameter
  • Hollow top finish with ribs for easy retrieval
Description & Specifications
Specifications
Fit tubes with 12 to 16mm diameter