3M™ DBI-Sala™ Rope Lifeline Assembly
With stainless-steel self-locking snap hooks at either end for easy attachment and set-up
Manufacturer:  3M™ 1202754C
 View more versions of this product
Catalog No. 19-066-836
Description
Description
  • Hooks to overhead anchor attachment point and is made to work with rope grab as part of fall protection system
  • Rope grab follows worker as he climbs and engages in the event of fall
  • Gates have 3/4 in. openings and are rated to 3,600 lb. each
  • Blended polyester/polypropylene rope
  • Easy to deploy and flake
  • Does not tend to knot or kink
  • UV-and abrasion-resistant
  • Suitable for creating fall protection system for workers at height and still needs mobility
Compliance
Each of snap hooks in this lifeline assembly meets OSHA and ANSI 359 Standards.
Specifications
Specifications
OSHA, ANSI, ANSI Z359
Self locking snap hook at one end
1632.93 kg
1.9 cm
9.14 m
Rope lifeline assembly
Rope grab
3600 lb.
0.75 in.
30 ft.
Polyester, Polypropylene