Please login to your online account to display your discounted pricing

Alfa Aesar™ Cantharidic acid, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67282MA

 View more versions of this product

Catalog No. AAJ67282MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 216° to 218°C
Formula Weight 214.22
Chemical Name or Material Cantharidic acid
CAS 28874-45-5
MDL Number MFCD00047137
Assay 98%
Color White
Quantity 10mg
Physical Form Solid

Specifications

Chemical Name or Material Cantharidic acid
CAS 28874-45-5
Formula Weight 214.22
MDL Number MFCD00047137