Please login to your online account to display your discounted pricing

Alfa Aesar™ Cantharidic acid, 98%

Chemical Identifiers

Chemical Name or Material Cantharidic acid
CAS 28874-45-5
Formula Weight 214.22
MDL Number MFCD00047137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67282MA

 
alfa aesar™
J67282MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 216° to 218°C
Formula Weight 214.22
Chemical Name or Material Cantharidic acid
CAS 28874-45-5
MDL Number MFCD00047137
Assay 98%
Color White
Quantity 10mg
Physical Form Solid