missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cannon™ Cannon™ Cold-Cranking Simulator Standards

Synthetic base oil standards used to calibrate the Cannon Cold-Cranking Simulator (CCS) oil testing. Cannon CL Standards for Cold-Cranking Simulator provides certified dynamic viscosity data (in cP or mPa/s) at temperatures from -5° to -35°C.

$340.91 - $348.00

Specifications

Quantity 500 mL
Type Viscosity Standard
For Use With (Equipment) Cannon Cold Cranking Simulator
Certifications/Compliance ASTM D 5293, SAE, NIST
For Use With (Application) Calibration
View More Specs

Products 2
Catalog Number Mfr. No. Viscosity Price Quantity  
Catalog Number Mfr. No. Viscosity Price Quantity  
23-023-722
View Documents Promotion Details
Cannon™
9727N30016
7300 cSt at -20°C
Each for $340.91
 
23-023-724
View Documents Promotion Details
Cannon™
9727N34016
21,000 cSt at -20°C
Each for $348.00
 
Description

Description

  • Conform to SAE specification J300 and ASTM D 5293
  • Viscosities based on NIST value of 1.0016 centipoise for water at 20°C
  • Traceable to NIST
  • Supplied in one pint bottle

Certifications

All standards are traceable to NIST™. NIST traceable

Specifications

Specifications

500 mL
Cannon Cold Cranking Simulator
Calibration
Viscosity Standard
ASTM D 5293, SAE, NIST
Liquid
SDS
missing translation for 'documents'

missing translation for 'documents'