Please login to your online account to display your discounted pricing

Candesartan cilexetil, Tocris™

Prodrug of candesartan, an AT1 receptor antagonist

$88.05 - $376.43

Chemical Identifiers

Chemical Name or Material Candesartan cilexetil
Assay Percent Range >99%
Molecular Formula C33H34N6O6
Formula Weight 610.66
Synonym 2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1, 1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

479250

 
r & d systems
479250
50mg Each for $376.43

479210

 
r & d systems
479210
10mg Each for $88.05
Description & Specifications

Specifications

Chemical Name or Material Candesartan cilexetil
Quantity 50mg
CAS 145040-37-5
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C33H34N6O6
Formula Weight 610.66
Synonym 2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1, 1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester
Safety and Handling

Store at +4°C