Please login to your online account to display your discounted pricing

Canada balsam, natural filtered, Alfa Aesar™

$446.00 - $446.00

Chemical Identifiers

Chemical Name or Material Canada balsam
CAS 8007-47-4
Name Note natural filtered
MDL Number MFCD00132800
UN Number UN1993
Merck Index 14,946
Synonym Balsam Canada
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1628936

 
alfa aesar
A1628936
Each for $446.00
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Canada balsam
TSCA TSCA
Name Note natural filtered
MDL Number MFCD00132800
CAS 8007-47-4
UN Number UN1993
Merck Index 14,946
Synonym Balsam Canada
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226;
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number CP2352500
Flash Point 43°C (109°F)

Applications
Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

Solubility
It is soluble in benzene, chloroform, xylene, ethyl acetate, ether, oil of cedar, and oil of turpentine. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226;
Flammable liquid and vapour.