missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Camptothecin, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 276721000

 View more versions of this product

Catalog No. AC276721000



Description

Description

Specifications

Specifications

(+)-Camptothecin
1.0% max.
Yellow
100mg
7689-03-4
100.0
C20H16N2O4
15, 1737
Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetonirile,dmso,,chloroform,hydrochloric acid
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
24360
348.35
98%
Authentic
Glass bottle
260.0°C
98%
97.5
97.5% min. (HPLC)
MFCD00081076
+-camptothecin, +-camptothecine, 20 s-camptothecine, 21,22-secocamptothecin-21-oic acid lactone, campathecin, camptothecin, camptothecine, d-camptothecin, s-+-camptothecin, s-camptothecin
VSJKWCGYPAHWDS-FQEVSTJZSA-N
348.35
CHEBI:27656
Powder

Chemical Identifiers

7689-03-4
348.35
VSJKWCGYPAHWDS-FQEVSTJZSA-N
24360
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
C20H16N2O4
MFCD00081076
+-camptothecin, +-camptothecine, 20 s-camptothecine, 21,22-secocamptothecin-21-oic acid lactone, campathecin, camptothecin, camptothecine, d-camptothecin, s-+-camptothecin, s-camptothecin
CHEBI:27656
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : UQ0492000

TSCA : TSCA

SDS Documents