Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Camphoric anhydride, 98%

$70.35 - $197.03

Chemical Identifiers

Chemical Name or Material (+/-)-Camphoric anhydride
CAS 595-30-2
Molecular Formula C10H14O3
Formula Weight 182.22
MDL Number MFCD00067307
Beilstein 83114
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2340114

  SDS
alfa aesar™
B2340114
25g Each for $70.35

AAB2340118

  SDS
alfa aesar™
B2340118
50g Each for $113.59

AAB2340122

  SDS
alfa aesar™
B2340122
100g Each for $197.03
Description & Specifications

Specifications

Formula Weight 182.22
Chemical Name or Material (+/-)-Camphoric anhydride
Assay 98%
Molecular Formula C10H14O3
MDL Number MFCD00067307
CAS 595-30-2
Beilstein 83114
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 269° to 271°C
Melting Point 220° to 225°C
Quantity 25g

Applications
(+/-)-Camphoric anhydride is useful in the synthesis of aliphatic-aromatic copolyesters. It is also used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture and oxidizing agents. Protect from humidity and water.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.