missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Camphene, 75%, remainder mainly alpha-fenchene, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 295570010

Catalog No. AC295570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Camphene
79-92-5
159.0°C to 160.0°C
Authentic
Glass bottle
MFCD00066603
05, 156
camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
CRPUJAZIXJMDBK-UHFFFAOYSA-N
3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
6616
CHEBI:3830
75%
75%
0.85
36°C
70% min. (GC)
C10H16
1L
15, 1732
Solubility in water: negligible. Other solubilities: soluble in ether,cyclohexane,cyclohexene,,dioxane and chloroform,slightly soluble in alcohol
CC1(C2CCC(C2)C1=C)C
136.24
1.6 mPa.s (50°C)
136.24
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Flammable solid.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If eye irritation persists: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiti

GHS Signal Word: Warning

EINECSNumber : 201-234-8

RTECSNumber : EX1055000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only