Learn More
(-)-Camphanic acid chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 209890010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chiral derivatization reagentSpecifications
(-)-Camphanic acid chloride | |
39637-74-6 | |
100.0 | |
98% | |
C10H13ClO3 | |
1g | |
1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride | |
PAXWODJTHKJQDZ-QVDQXJPCSA-N | |
(4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride | |
12364938 | |
98% |
98% | |
97.5 | |
Authentic | |
Plastic bottle | |
MFCD00135626 | |
− 18.00 | |
Solubility in water: decomposes | |
CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C | |
216.66 | |
216.66 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 254-552-4
TSCA : TSCA
RUO â Research Use Only