missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cAMP Parameter Assay Kit, R&D Systems

$415.83 - $1,995.75

Specifications

Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA kit
For Use With (Application) ELISA
Sensitivity 8.57pmol/mL
Species Multi-species
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sufficient For Quantity Price Quantity  
Catalog Number Mfr. No. Sufficient For Quantity Price Quantity  
KGE002B R & D Systems
RDSKGE002B
1 x 96-well plate 1 kit Each for $415.83
SKGE002B R & D Systems
RDSSKGE002B
6 x 96-well plates 1 Pk. Each for $1,995.75
Specifications

Description

Adenosine 3',5'-cyclic monophosphate (cAMP) is one of the most important intracellular second messengers. Several hormones are known to activate cAMP and subsequent signaling cascades via their indirect influence on adenylate cyclase, the enzyme responsible for the conversion of ATP to cAMP.

Assay Type: Solid Phase Competitive ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Cell Lysates (100μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL), Saliva (50μL), Urine (10μL)
Sensitivity: 8.57 pmol/mL
Specificity: Measures cyclic AMP levels in various samples
Cross Reactivity: Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
ELISA
Multi-species
ELISA kit
8.57pmol/mL