Learn More
Thermo Scientific Chemicals cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
For cellular and molecular biology applications
Supplier: Thermo Scientific Chemicals J66438MCR
Description
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) acts as inhibitor of c-AMP-dependent protein kinase These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) | |
RUO - research use only |
-30°C to -10°C | |
2222.40 | |
1 mg | |
Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp |
RUO – Research Use Only