Please login to your online account to display your discounted pricing

Camostat mesylate, Tocris

Orally active protease inhibitor

$76.90 - $277.92

Chemical Identifiers

Chemical Name or Material Camostat mesylate
Assay Percent Range >98%
Molecular Formula C20H22N4O5·CH3SO3H
Formula Weight 494.52
Synonym 4-[[4-[(Aminoiminomethyl)amino]benzoyl]oxy]benzeneacetic acid 2-(dimethylamino)-2-oxoethyl ester methanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

319310

 
r & d systems
RDS319310
10mg Each for $76.90

319350

 
r & d systems
RDS319350
50mg Each for $277.92
Description & Specifications

Specifications

Formula Weight 494.52
Quantity 10mg
Chemical Name or Material Camostat mesylate
Assay Percent Range >98%
Molecular Formula C20H22N4O5·CH3SO3H
CAS 59721-29-8
Synonym 4-[[4-[(Aminoiminomethyl)amino]benzoyl]oxy]benzeneacetic acid 2-(dimethylamino)-2-oxoethyl ester methanesulfonate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature