missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ CAMKK2 (CaMKK Beta) Recombinant Human Protein
CAMKK2 (CaMKK beta) is a serine/threonine kinase.
$4085.00 - $19320.00
Specifications
Accession Number | NP_757380.1 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, and 50% Glycerol |
Molecular Weight (g/mol) | 87.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7777A
|
Invitrogen™
PR7777A |
100μg |
Each for $4,085.00
|
|
|||||
PV4322
|
Invitrogen™
PV4322 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CAMKK2 (CaMKK beta) is involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.
Specifications
NP_757380.1 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, and 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
87.1kDa | |
RUO - research use only | |
CAMKK2 (CaMKK Beta) | |
Human | |
GST Tag | |
Frozen |