Invitrogen™ CAMKK2 (CaMKK Beta) Recombinant Human Protein DFS Item
CAMKK2 (CaMKK beta) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4322
 View more versions of this product
Catalog No. PV4322
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
CAMKK2 (CaMKK beta) is involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.
Specifications
Specifications
NP_757380.1
Frozen
87.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CAMKK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, and 50% Glycerol
1mg
RUO - research use only
CAMKK2 (CaMKK Beta)
Baculovirus
Human
GST Tag
Documents