missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ CAMKK1 (CAMKKA) Recombinant Human Protein
CAMKK1 (CAMKKA) is a serine/threonine protein kinase family member.
Manufacturer: Invitrogen™ LSPV4671
Description
CAMKK1 (CAMKKA) belongs to the Ca2+/calmodulin-dependent protein kinase subfamily.
Specifications
NP_115670 | |
Frozen | |
59.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
Kinase Assay | |
CAMKK Kinase Family | |
Yes |
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma | |
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol | |
1mg | |
RUO - research use only | |
CAMKK1 (CAMKKA) | |
Baculovirus | |
Human | |
His Tag |