Invitrogen™ CAMKK1 (CAMKKA) Recombinant Human Protein DFS Item
CAMKK1 (CAMKKA) is a serine/threonine protein kinase family member.
Manufacturer:  Invitrogen™ LSPV4670
 View more versions of this product
Catalog No. PV4670
Description
Description
CAMKK1 (CAMKKA) belongs to the Ca2+/calmodulin-dependent protein kinase subfamily.
Specifications
Specifications
NP_115670
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol
10μg
RUO - research use only
Serine/Threonine Kinases
calcium/calmodulin-dependent protein kinase kinase 1, alpha
Kinases & Inhibitors
CAMKK Kinase Family
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
59.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length, Recombinant
CAMKK1 (CAMKKA)
Kinase Assay
Baculovirus
Human
His Tag
Documents