Invitrogen™ CAMK2G (CaMKII gamma) Recombinant Human Protein DFS Item
CAMK2G (CaMKII gamma) is a ubiquitous serine/threonine protein kinase that has been implicated in diverse effects of hormones and neurotransmitters. Recombinant human CAMK2G (c-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
Manufacturer:  Invitrogen™ LSPV6270
 View more versions of this product
Catalog No. PV6270
Description
Description
CAMK2G has six alternatively spliced variants that encode six different isoforms. Some of these variants have been identified in human tumors. Transgenic mice expressing a partially calcium-independent mutant form of CAMKG showed 1.5- to 2-fold increase in the thymus of these mice, at least in part due to an increase in the life span of double-positive thymocytes. There was an increase in the number of T cells in the secondary lymphoid organs that had acquired an antigen-dependent memory phenotype.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
60.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
aa c-terminus. Contains S36P SNP
Yes
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
1mg
Kinase Assay
Human
GST Tag
Documents