missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CAMK2A (CaMKII Alpha) Recombinant Human Protein DFS Item

Manufacturer:  Life Technologies LSPV3142

 View more versions of this product

Catalog No. PV3142



Description

Description

CAMK2A (CaMKII alpha) is a serine⁄threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity.
Specifications

Specifications

NP_741960
Storage Buffer: 20 mM Tris pH 7.5, 100 mM NaCl, 0.5 mM EDTA, 0.05% NP-40, 2 mM DTT and 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
CAMK2A
Baculovirus
Full Length, Recombinant
Signal Transduction
Kinase Assay
56.8 kDa
CAMK2 Kinase Family
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Ser⁄Thr Kinases: CAMK Group
LanthaScreen® Antibody, LanthaScreen® Substrate, Z'-LYTE™, Omnia® Bulk Substrate
calcium⁄calmodulin-dependent protein kinase (CaM kinase) II alpha
Kinases & Inhibitors
Serine⁄Threonine Kinases
Human
25400, 815, 816, 817, 818
CAMK2A (CaMKII Alpha)
His Tag
Documents