Invitrogen™ CAMK1D (CaMKI Delta) Recombinant Human Protein DFS Item
CAMK1D (CaMK1 delta) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family.
Manufacturer:  Invitrogen™ LSPV3977
 View more versions of this product
Catalog No. PV3977
Description
Description
CAMK1D is expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis.
Specifications
Specifications
NP_705718
Frozen
47kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CAMK1 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
CAMK1D (CaMKI Delta)
Baculovirus
Human
His Tag
Documents