Please login to your online account to display your discounted pricing

Cambridge Diagnostic Products, Inc.™ Camco™ Wright Stain, Fixative and Buffer


Plastic container is easy to store and allows convenient dispensing into slide stainer traps

Manufacturer: cambridge diagnostic  CAM402

 View more versions of this product

Catalog No. 04-330-5

  • / Each
 

Description & Specifications

Specifications

Quantity 10 × 4.4g vial
For Use With (Application) For in-vitro diagnostic use
pH 7.15
Storage Requirements 10° to 30°C
Type Buffer