Cambridge Diagnostic Products, Inc.™ Camco™ Stain Pak
Manufacturer:  Cambridge Diagnostic Products, Inc.™ CAM702A
 View more versions of this product
Catalog No. 04-330-8
Description
Description
  • Provides rapid, consistent differential staining of blood smears approximating results obtained from Wright-Giemsa staining
  • In addition to highly purified dyestuff and buffers, CAMCO Stain Pak contains important electrolytes that enhance the staining of the nuclei and improve contrast
  • Fixative solution is used to stabilize the cell and its contents
  • Convenient sizes available
Order Info
Stains are packaged in one-gallon bottles except for Stain Pak, which is packaged in one 473mL bottle each of Fixative, Solution I, and Solution II
Specifications
Specifications
1 gal.
Liquid
For rapid differential staining of Blood smears
64°C
Fixative, 1 gal.
Clear
Documents