Please login to your online account to display your discounted pricing

Cambridge Diagnostic Products, Inc.™ Camco™ Stain Pak


Manufacturer: cambridge diagnostic  CAM702

 View more versions of this product

Catalog No. 04-330-7

  • / Pack of 3 Each
 

Description & Specifications

Specifications

Quantity 3 x 473mL
For Use With (Application) For rapid differential staining of Blood smears
Boiling Point 100°C
Format Liquid
Type Stain Pak