Please login to your online account to display your discounted pricing

Cambridge Diagnostic Products, Inc.™ Camco™ Stain Pak


Manufacturer: cambridge diagnostic  CAM702B

 View more versions of this product

Catalog No. 04-330-9

  • / Each
 

Description & Specifications

Specifications

Type Solution I (Red), 1 gal.
Quantity 1 gal.
For Use With (Application) For rapid differential staining of Blood smears
Boiling Point 100°C
Format Liquid