Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Calretinin Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calretinin Ab-2, Mouse Monoclonal Antibody.

$306.02 - $1,504.14

Specifications

Antigen Calretinin Ab-2
Clone DAK Calret 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1859R7

 
thermo scientific™ lab vision™
MS1859R7
7mL Each for $1,087.14

MS1859S

 
thermo scientific™ lab vision™
MS1859S
1mL Each for $1,504.14

MS1859S0

 
thermo scientific™ lab vision™
MS1859S0
100µL Each for $306.02

MS1859S1

 
thermo scientific™ lab vision™
MS1859S1
500µL Each for $1,055.12
Description & Specifications

Specifications

Antigen Calretinin Ab-2
Clone DAK Calret 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Recombinant full length human calretinin protein cloned from Mero-41 cells

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calretinin Ab-2, Mouse Monoclonal Antibody.

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Mouse

Clone: DAK Calret 1

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human calretinin protein cloned from Mero-41 cells

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO