Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Calponin-1, Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ Calponin-1, Rabbit Monoclonal Antibody.

$332.57 - $1,926.08

Specifications

Antigen Calponin
Clone EP798Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RM2102R7

 
thermo scientific™ lab vision™
RM2102R7
7mL Each for $831.43

RM2102S

 
thermo scientific™ lab vision™
RM2102S
1mL Each for $1,926.08

RM2102S0

 
thermo scientific™ lab vision™
RM2102S0
100µL Each for $332.57
Description & Specifications

Specifications

Antigen Calponin
Clone EP798Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen A synthetic peptide derived from near the C-terminus of human Calponin-1

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ Calponin-1, Rabbit Monoclonal Antibody.  Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Calponin, a thin filament associated protein, is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.

Host Species: Rabbit

Clone: EP798Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the C-terminus of human Calponin-1

Molecular Weight: 29/34 kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasm Myoepithelial cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)