Please login to your online account to display your discounted pricing

CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T.Baker™


Suitable for Use in Hemodialysis

Quantity 2.5kg
Packaging Wide Mouth PVC Coated Glass Bottle

Manufacturer: avantor performance  133605

 View more versions of this product

Catalog No. 02002519

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/USP
Formula Weight 147.01
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
CAS 10035-04-8
Packaging Wide Mouth PVC Coated Glass Bottle
Quantity 2.5kg
Specific Gravity 1L = 0.84kg
Molecular Formula CaCl2·2H2O

Specifications

Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
CAS 10035-04-8
Purity Grade Multi-Compendial/USP
Molecular Formula CaCl2·2H2O
Formula Weight 147.01