Please login to your online account to display your discounted pricing

Calpeptin, Tocris


Calpain and cathepsin L inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS044810

 View more versions of this product

Catalog No. 044810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 362.47
Molecular Formula C20H30N2O4
Assay Percent Range >98%
Synonym N-Benzyloxycarbonyl-L-leucylnorleucinal
CAS 117591-20-5
Chemical Name or Material Calpeptin
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Calpeptin
Assay Percent Range >98%
Molecular Formula C20H30N2O4
Formula Weight 362.47
Synonym N-Benzyloxycarbonyl-L-leucylnorleucinal
Accessories

Desiccate at +4°C