missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calpeptin, Tocris

Calpain and cathepsin L inhibitor

Manufacturer:  R & D Systems RDS044850

 View more versions of this product

Catalog No. 044850


 

Specifications Description & Specifications

Specifications

Calpeptin
117591-20-5
C20H30N2O4
N-Benzyloxycarbonyl-L-leucylnorleucinal
50mg
>98%
362.47

Chemical Identifiers

>98%
362.47
Calpeptin
C20H30N2O4
N-Benzyloxycarbonyl-L-leucylnorleucinal

Safety and Handling

Recommended Storage : Desiccate at +4°C