Please login to your online account to display your discounted pricing

Alfa Aesar™ Caloxin 2A1

For cellular and molecular biology applications

Specifications

Product Type Caloxin 2A1
Molecular Weight 1478.54
Storage Requirements -30° to -10°C
Sequence Val-Ser-Asn-Ser-Asn-Trp-Pro-Ser-Phe-Pro-Ser-Ser-Gly-Gly-Gly-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66428EXD

 
alfa aesar™
J66428EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66428MCR

 
alfa aesar™
J66428MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Caloxin 2A1
Molecular Weight 1478.54
Storage Requirements -30° to -10°C
Sequence Val-Ser-Asn-Ser-Asn-Trp-Pro-Ser-Phe-Pro-Ser-Ser-Gly-Gly-Gly-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC