Please login to your online account to display your discounted pricing
Dräger™ Calibration Gases
$264.48 - $504.62
Specifications
Product Type Calibration Gas
Shelf Life 36 Months
Volume (English) Gas 27.2 gal.
View More Specs
Products
Catalog Number Mfr. No. Concentration Pressure Gas Type Volume (Metric) Gas Price Quantity    
17-987-48L
dräger™
4595194
1000ppm 1000psi Carbon Dioxide 103L Each for $266.15
17-986-5A
dräger™
4502155
25ppm 500psi Hydrogen Sulfide 58L Each for $305.68
19-038-746
dräger™
4594943
LEL, methane 50%, carbon monoxide 100ppm, hydrogen sulfide 25ppm, oxygen 20.9% 500psi Methane, Carbon Monoxide, Hydrogen Sulfide, Oxygen 58L Each for $504.62
19-121-382
dräger™
4594947
30% LEL, carbon monoxide 100, oxygen 17% 1000psi C5H12, Carbon Monoxide, Oxygen 103L Each for $338.32
19-130-5826
dräger™
4520128
100ppm 1000psi Carbon Monoxide 103L Each for $264.48
19-130-5827
dräger™
4594655
LEL, methane 50%, carbon monoxide 100ppm, hydrogen sulfide 25ppm, oxygen 17% 500psi Methane, Carbon Monoxide, Hydrogen Sulfide, Oxygen 58L Each for $504.62
19-815-812
dräger™
4594960
10ppm 500psi Hydrogen Sulfide 58L Each for $303.91
19-815-811
dräger™
4594645
100ppm 1000psi Ethylene 103L Each for $440.81
Description & Specifications
Specifications
Product Type Calibration Gas
Shelf Life 36 Months
Volume (English) Gas 27.2 gal.
Safety and Handling
36 Months