H-B Frio-Temp SP Scienceware™ Calibrated Electronic Verification Thermometers, Calibrated for Refrigerators PROMO
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Available calibrated for specific applications and equipment.
$96.00 - $115.50
Specifications
Calibration Points 4°C Refrigerator Zone 1 External
Calibrated Yes
Resolution 0.1°C/°F
Includes Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Certifications/Compliance Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
View More Specs
Products 3
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Fill Probe Type Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Fill Probe Type Price Quantity  
13202771PM View Documents H-B Frio-Temp
B602100100PM
−50° to 300°C (−58° to572°F) Glass Bottle/Glass Beads Angled stainless steel Each for $114.00
13202776PM View Documents H-B Frio-Temp
B602100600PM
−50° to 70°C (−58° to158°F) External; 0° to50°C (32° to122°F) Internal Plastic Bottle/Organic Liquid Waterproof Each for $115.50
13202780PM View Documents H-B Frio-Temp
B602101000PM
−50° to 70°C (−58° to158°F) External; −10° to50°C (14° to122°F) Internal Plastic Bottle/Glass Beads Waterproof Each for $96.00
Description
Description
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No. 2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE marked
Specifications
Specifications
4°C Refrigerator Zone 1 External
0.1°C/°F
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
Yes
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)