H-B Frio-Temp SP Scienceware™ Calibrated Electronic Verification Thermometers, Calibrated for Incubators PROMO
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Available calibrated for specific applications and equipment.
$96.00 - $126.75
Specifications
Calibration Points 37°C Incubator
Calibrated Incubator
For Use With (Application) Temperature Calibration and Verification
Includes Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Certifications/Compliance Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
View More Specs
Products 4
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Fill Height (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Fill Height (Metric) Width (Metric) Price Quantity  
13202772PM View Documents H-B Frio-Temp
B602100200PM
−50° to 300°C Glass Bottle/Glass Beads 95mm 64mm Each for $114.00
13202774PM View Documents H-B Frio-Temp
B602100400PM
−40° to 70°C External Plastic Bottle (2)/Glass Beads 19mm 40mm Each for $126.75
13202777PM View Documents H-B Frio-Temp
B602100700PM
−50° to 70°C External; 0° to 50°C Internal Plastic Bottle/Organic Liquid 97mm 65mm Each for $115.50
13202781PM View Documents H-B Frio-Temp
B602101100PM
−50° to 70°C External; −10° to 50°C Internal Plastic Bottle/Glass Beads 44mm 90mm Each for $96.00
Description
Description
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No. 2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE marked
Specifications
Specifications
37°C Incubator
Temperature Calibration and Verification
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
Incubator
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)