H-B Frio-Temp SP Scienceware™ Calibrated Electronic Verification Thermometers, Calibrated for General Applications PROMO
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Available calibrated for specific applications and equipment.
$60.75 - $99.00
Specifications
Resolution 0.1°C/°F
Calibrated General
Includes Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Certifications/Compliance Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
For Use With (Application) Temperature Calibration and Verification
View More Specs
Products 6
Catalog Number Mfr. No. Temperature Range (Metric) Calibration Points Bottle Fill Probe Type Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Calibration Points Bottle Fill Probe Type Price Quantity  
13201457PM View Documents H-B Frio-Temp
B610000000PM
−50° to 200°C 0°, 100°C Glass Bottle/Glass Beads Angled stainless steel Each for $70.50
13201458PM View Documents H-B Frio-Temp
B610000200PM
−50° to 70°C External;−10° to 50°C Internal 0°, 50°C Plastic Bottle/Glass Beads Waterproof Each for $60.75
13201460PM View Documents H-B Frio-Temp
B610000400PM
−50° to 70°C External; 0° to 50°C Internal 0°, 50°C Plastic Bottle/Organic Liquid Waterproof Each for $81.00
13201861PM View Documents H-B Frio-Temp
B610005000PM
−50° to 200°C 0°, 100°C Glass Bottle/Glass Beads Angled stainless steel Each for $76.45
13202784PM View Documents H-B Frio-Temp
B610000600PM
−40° to 70°C External 0°, 50°C Plastic Bottle (2)/Organic Liquid Waterproof Each for $69.00
13203029PM View Documents H-B Frio-Temp
B610000100RMDPM
−50° to 300°C 0°, 100°C Glass Bottle/Glass Beads Angled Stainless Steel Each for $99.00
Description
Description
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No. 2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE marked
Specifications
Specifications
0.1°C/°F
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Temperature Calibration and Verification
General
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.