Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Caldesmon, LMW Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Manufacturer: thermo scientific™ lab vision™  MS1251P1ABX

 View more versions of this product

Catalog No. MS1251P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Caldesmon, LMW Ab-2
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone l-CALD
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Recombinant caldesmon
Isotype IgG2a kappa
Purification Method Rh30 cells, uterus
Quantity 100μL
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Caldesmon, LMW Ab-2, Mouse Monoclonal Antibody.

Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.

Host Species: Mouse

Clone: l-CALD

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant caldesmon

Molecular Weight: 80kDa (l-caldesmon)

Positive Control: Rh30 cells, Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO