Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Caldesmon, LMW Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Manufacturer: thermo scientific™ lab vision™  MS1251PABX

 View more versions of this product

Catalog No. MS1251PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG2a kappa
Immunogen Recombinant caldesmon
Antigen Caldesmon, LMW Ab-2
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Clone l-CALD
Quantity 200µL
Format Concentrated
Conjugate Unlabeled
Type Primary
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Muscle Biology
Classification Monoclonal
Purification Method Rh30 cells, uterus

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Caldesmon, LMW Ab-2, Mouse Monoclonal Antibody.

Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.

Host Species: Mouse

Clone: l-CALD

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant caldesmon

Molecular Weight: 80kDa (l-caldesmon)

Positive Control: Rh30 cells, Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO