missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calconcarboxylic acid, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2345909

 View more versions of this product

Catalog No. AAB2345909



Description

Description

Calconcarboxylic acid is used as an indicator for calcium titrations with ethylenediaminetetraacetic acid (EDTA) in the presence of magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calconcarboxylic acid is used as an indicator for calcium titrations with ethylenediaminetetraacetic acid (EDTA) in the presence of magnesium.

Solubility
Soluble in water, ethanol, sodium hydroxide and methanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Calconcarboxylic acid
∼300°C (decomposition)
MFCD00004078
727492
Soluble in water,ethanol,sodium hydroxide and methanol.
C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O
438.41
438.42
3737-95-9
C21H14N2O7S
10 g
calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nn
ULIVOAKVRBXKKS-PYCFMQQDSA-N
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
5895210

Chemical Identifiers

3737-95-9
438.41
ULIVOAKVRBXKKS-PYCFMQQDSA-N
5895210
C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O
C21H14N2O7S
MFCD00004078
calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nn
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-117-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only