Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium zirconium isopropoxide, 10% w/v in isopropanol

Precursor for mixed Ca, Zr oxide by Sol-Gel method

Chemical Identifiers

Chemical Name or Material Calcium zirconium isopropoxide
CAS 334512-82-2
Linear Formula CaZr[OCH(CH3)2]6
Formula Weight 485.83
MDL Number MFCD01865977
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41971AC

 
alfa aesar™
41971AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41971AE

 
alfa aesar™
41971AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 485.83
Health Hazard 3 GHS P Statement
P210-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Linear Formula CaZr[OCH(CH3)2]6
MDL Number MFCD01865977
Health Hazard 2 GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Chemical Name or Material Calcium zirconium isopropoxide
Physical Form Liquid
CAS 334512-82-2
UN Number UN1219
Health Hazard 1 Danger
Color Yellow
Concentration 10% w/v in isopropanol
Quantity 25mL
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Safety and Handling

Danger

GHS P Statement
P210-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.