missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium turnings, 98% (metals basis), Alfa Aesar

Manufacturer:  Alfa Aesar 1012509

 View more versions of this product

Catalog No. AA1012509



Specifications

Specifications

Calcium
7440-70-2
98% (metals basis)
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
[Ca]
40.078
CHEBI:29320
Turnings
10g
1.54
Ca
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
5460341
40.078

Chemical Identifiers

7440-70-2
40.078
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
CHEBI:29320
[Ca]
Ca
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Documents