missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium trifluoromethanesulfonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC358680100

 View more versions of this product

Catalog No. AC358680100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Powder
10g
98%
Authentic
98%
C2CaF6O6S2
Grayish
338.21
(F3CSO3)2Ca
Glass bottle
Calcium trifluoromethanesulfonate
55120-75-7
MFCD00143911

Chemical Identifiers

55120-75-7
C2CaF6O6S2
338.21
Calcium trifluoromethanesulfonate, 98%
98%
(F3CSO3)2Ca
MFCD00143911
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.