Please login to your online account to display your discounted pricing

Calcium trifluoromethanesulfonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Calcium trifluoromethanesulfonate
Name Note 98%
CAS 55120-75-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2CaF6O6S2
Linear Formula (F3CSO3)2Ca
Formula Weight 338.21
MDL Number MFCD00143911
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC358680100

  SDS
thermo fisher scientific
AC358680100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 338.21
Color Grayish
Quantity 10g
Chemical Name or Material Calcium trifluoromethanesulfonate
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C2CaF6O6S2
Linear Formula (F3CSO3)2Ca
CAS 55120-75-7
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00143911
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.