missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium thiosulfate, pure, 30-50% solution in water, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC447870010

 View more versions of this product

Catalog No. AC447870010



Specifications

Specifications

Calcium thiosulfate
CaO3S2
calcium hyposulfite, calcium thiosulfate, calcium thiosulfate ca s2o3, calcium thiosulphate, caswell no. 153, epa pesticide chemical code 080101, tecesal, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1, unii-2io10o728e
FAYYUXPSKDFLEC-UHFFFAOYSA-L
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
24964
Pure
Glass bottle
1L
10124-41-1,7732-18-5
CaS2O3
Solubility in water: soluble
[O-]S(=O)(=S)[O-].[Ca+2]
152.21
152.21
1.2450g/mL
1.245

Chemical Identifiers

10124-41-1
152.21
calcium hyposulfite, calcium thiosulfate, calcium thiosulfate ca s2o3, calcium thiosulphate, caswell no. 153, epa pesticide chemical code 080101, tecesal, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1, unii-2io10o728e
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
CaO3S2
FAYYUXPSKDFLEC-UHFFFAOYSA-L
24964
[O-]S(=O)(=S)[O-].[Ca+2]
Safety and Handling

Safety and Handling

EINECSNumber : 233-333-7

SDS Documents