Learn More
Calcium thioglycolate trihydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 367035000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium thioglycolate trihydrate | |
270°C to 280°C | |
Authentic | |
Plastic bottle | |
MFCD00150707 | |
15, 1713 | |
Solubility in water: 70g/L (25°C). Other solubilities: very slightly soluble in alcohol,chloroform,practical insoluble in ether,petroleum ether,,benzene | |
O.O.O.[Ca].OC(=O)CS | |
186.24 | |
184.23 | |
Powder |
65208-41-5 | |
White | |
98.5% min. (Iodometry) | |
C2H10CaO5S | |
500 g | |
calciumthioglycolatetrihydrate | |
GDECCIJZCWLFSB-UHFFFAOYSA-N | |
2-sulfanylacetic acid calcium trihydrate | |
131881455 | |
99% |
Chemical Identifiers
65208-41-5 | |
186.24 | |
GDECCIJZCWLFSB-UHFFFAOYSA-N | |
131881455 |
C2H10CaO5S | |
MFCD00150707 | |
calciumthioglycolatetrihydrate | |
O.O.O.[Ca].OC(=O)CS |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
RUO – Research Use Only