missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%

Manufacturer:  Alfa Aesar™ 4364614

 View more versions of this product

Catalog No. AA4364614



Specifications

Specifications

Calcium sulfide
(metals basis excluding Mg), Mg <1%
MFCD00015990
14,1707
S=[Ca]
72.138
72.14
99%
−100 Mesh
Moisture Sensitive
25g
20548-54-3
CaS
UN3134
JGIATAMCQXIDNZ-UHFFFAOYSA-N
sulfanylidenecalcium
30182
Powder
2.5g/mL
Stench
White

Chemical Identifiers

20548-54-3
72.138
JGIATAMCQXIDNZ-UHFFFAOYSA-N
sulfanylidenecalcium
CaS
MFCD00015990
30182
S=[Ca]
Safety and Handling

Safety and Handling

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P231+P232-P261-P280-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.

TSCA : TSCA

Documents