Learn More
Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01312114
Description
It is used as luminous paint, depilatory, lubricant additive, ore dressing and flotation agent and as an arsenic-free hydrogen sulfide. It is also used as a depilatory in the tanning industry and in cosmetics, in homeopathy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium sulfide | |
20548-54-3 | |
25 g | |
CaS | |
UN3134 | |
Slightly soluble in water. Insoluble in alcohol | |
[S--].[Ca++] | |
72.14 | |
72.14 | |
Stench | |
2.5 g/mL |
(decomposition) | |
Powder | |
(metals basis excluding Sr), Sr typically 500ppm | |
MFCD00015990 | |
14,1707 | |
AGVJBLHVMNHENQ-UHFFFAOYSA-N | |
calcium sulfanediide | |
30182 | |
99.9% | |
Moisture sensitive |
Chemical Identifiers
20548-54-3 | |
72.14 | |
AGVJBLHVMNHENQ-UHFFFAOYSA-N | |
calcium sulfanediide |
CaS | |
MFCD00015990 | |
30182 | |
[S--].[Ca++] |
Safety and Handling
GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 243-873-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only