Please login to your online account to display your discounted pricing

Calcium Sulfate TS, Saturated Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Calcium Sulfate TS
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 96.0
CAS 10101-41-4
CAS Min % 5.49
CAS Max % 5.71
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

18235-16

 
ricca chemical company
RI1823.516
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

18235-4

 
ricca chemical company
RI1823.54
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Calcium Sulfate TS
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) Saturated Aqueous Solution
Shelf Life 12 months
CAS Min % 93.0
CAS Max % 96.0
CAS 10101-41-4
CAS Min % 5.49
CAS Max % 5.71
Packaging Poly natural
Safety and Handling

12 months