missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Dihydrate, ACS, 98-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C116112KG
Specifications
| 10101-41-4 | |
| 12 kg | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium dihydrate sulfate | |
| 98 to 102% | |
| Poly Pail |
| 1 | |
| CaH4O6S | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| ACS |
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| calcium dihydrate sulfate |
| CaH4O6S | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.