missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium sulfate dihydrate, 98+%, extra pure, ACROS Organics™
Manufacturer: Fisher Scientific Acros AC446380010
Specifications
Calcium sulfate dihydrate | |
98% min. (Complexometry) | |
CaSO4·2H2O | |
Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol,practically insoluble in most common organic,solvents | |
O.O.[O-]S(=O)(=O)[O-].[Ca+2] | |
172.17 | |
CHEBI:32583 | |
98+% | |
Plastic bottle | |
1kg |
10101-41-4 | |
CaO4S·2H2O | |
annaline, calcium sulfate dihydrate, compactrol, gips, gypsite, gypsum stone, landplaster, phosphogypsum, primoplast, satinite | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
calcium;sulfate;dihydrate | |
24928 | |
172.17 | |
Extra Pure | |
Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents |
Chemical Identifiers
10101-41-4 | |
172.17 | |
annaline, calcium sulfate dihydrate, compactrol, gips, gypsite, gypsum stone, landplaster, phosphogypsum, primoplast, satinite | |
CHEBI:32583 | |
O.O.[O-]S(=O)(=O)[O-].[Ca+2] |
CaO4S·2H2O | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
24928 | |
calcium;sulfate;dihydrate |
Safety and Handling