missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 98+%, for analysis

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol

Supplier:  Thermo Scientific Chemicals 225270025

 View more versions of this product

Catalog No. AC225270025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium sulfate dihydrate
10101-41-4
2.5 kg
CaH4O6S
MFCD00149625
14,1706
Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
O.O.[Ca++].[O-]S([O-])(=O)=O
172.16
CHEBI:32583
≥98%
Plastic bottle
White
Powder
98.0 to 102% (Complexometry)
CaSO4·2H2O
01,107
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PASHVRUKOFIRIK-UHFFFAOYSA-L
calcium;sulfate;dihydrate
24928
172.17
Analytical

Chemical Identifiers

10101-41-4
172.16
PASHVRUKOFIRIK-UHFFFAOYSA-L
24928
calcium;sulfate;dihydrate
CaH4O6S
MFCD00149625
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
CHEBI:32583
O.O.[Ca++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only